missing translation for 'onlineSavingsMsg'
Learn More

N-Succinimidyl Bromoacetate 98.0+%, TCI America™

Catalog No. s0852100mg
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S0852100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S0852100MG Supplier TCI America Supplier No. S0852100MG

Chemical Identifiers

CAS 42014-51-7
Molecular Formula C6H6BrNO4
Molecular Weight (g/mol) 236.021
MDL Number MFCD00058571
InChI Key NKUZQMZWTZAPSN-UHFFFAOYSA-N
Synonym n-succinimidyl bromoacetate, bromoacetic acid n-hydroxysuccinimide ester, 1-bromoacetyl oxy pyrrolidine-2,5-dione, 2,5-dioxoazolidinyl 2-bromoacetate, bromoacetic acid n-succinimidyl ester, pubchem11830, n-succinimidylbromoacetate, n-bromoacetoxy succinimide, bicl212, n-hydroxysuccinimide bromoacetate
PubChem CID 3565210
IUPAC Name (2,5-dioxopyrrolidin-1-yl) 2-bromoacetate
SMILES C1CC(=O)N(C1=O)OC(=O)CBr

Specifications

Melting Point 111°C
Color White-Yellow
Quantity 100 mg
Formula Weight 236.02
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N-Succinimidyl Bromoacetate
TSCA No
Recommended Storage Freezer