missing translation for 'onlineSavingsMsg'
Learn More

N-Succinimidyl Acetate 98.0+%, TCI America™

Catalog No. S08785G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S08785G 5 g
S087825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S08785G Supplier TCI America Supplier No. S08785G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 14464-29-0
Molecular Formula C6H7NO4
Molecular Weight (g/mol) 157.125
MDL Number MFCD00037916
InChI Key SIFCHNIAAPMMKG-UHFFFAOYSA-N
Synonym Acetic Acid N-Succinimidyl Ester, N-Acetoxysuccinimide
PubChem CID 84460
IUPAC Name (2,5-dioxopyrrolidin-1-yl) acetate
SMILES CC(=O)ON1C(=O)CCC1=O

Specifications

Melting Point 134°C
Color White
Quantity 5 g
Formula Weight 157.13
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N-Succinimidyl Acetate
TSCA No
Recommended Storage Refrigerator