missing translation for 'onlineSavingsMsg'
Learn More

N-Succinimidyl 4-(N-Maleimidomethyl)cyclohexanecarboxylate 98.0+%, TCI America™

Catalog No. p-7135965
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S0853100MG 100 mg
S08531G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S0853100MG Supplier TCI America Supplier No. S0853100MG
Only null left

Chemical Identifiers

CAS 64987-85-5
Molecular Formula C16H18N2O6
Molecular Weight (g/mol) 334.328
MDL Number MFCD00009634
InChI Key JJAHTWIKCUJRDK-UHFFFAOYSA-N
Synonym smcc, 2,5-dioxopyrrolidin-1-yl 4-2,5-dioxo-2,5-dihydro-1h-pyrrol-1-yl methyl cyclohexanecarboxylate, n-succinimidyl 4-n-maleimidomethyl cyclohexane-1-carboxylate, trans-4-maleimidomethyl cyclohexanecarboxylic acid-nhs, unii-b357p1g1if, n-succinimidyl 4-maleimidomethyl cyclohexane-1-carboxylate, n-4-carboxycyclohexylmethyl maleimide n-hydroxysuccinimide ester, trans-2,5-dioxopyrrolidin-1-yl 4-2,5-dioxo-2,5-dihydro-1h-pyrrol-1-yl methyl cyclohexanecarboxylate, n-hydroxysuccinimidyl 4-n-maleimidomethylcyclohexane-1-carboxylate, succinimidyl 4-n-maleimidomethyl cyclohexane-1-carboxylate
PubChem CID 125175
ChEBI CHEBI:63174
IUPAC Name (2,5-dioxopyrrolidin-1-yl) 4-[(2,5-dioxopyrrol-1-yl)methyl]cyclohexane-1-carboxylate
SMILES C1CC(CCC1CN2C(=O)C=CC2=O)C(=O)ON3C(=O)CCC3=O

Specifications

Melting Point 177°C
Color White
Quantity 100 mg
Formula Weight 334.33
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material N-Succinimidyl 4-(N-Maleimidomethyl)cyclohexanecarboxylate
TSCA No
Recommended Storage Freezer