missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Succinimidyl 3-Maleimidopropionate 98.0+%, TCI America™
Supplier: TCI America S04271G
Specifications
| N-Succinimidyl 3-Maleimidopropionate [Cross-linking Reagent] | |
| 168°C | |
| C11H10N2O6 | |
| 1 g | |
| JKHVDAUOODACDU-UHFFFAOYSA-N | |
| (2,5-dioxopyrrolidin-1-yl) 3-(2,5-dioxopyrrol-1-yl)propanoate | |
| 4620597 | |
| ≥98.0% (HPLC,N) |
| 55750-62-4 | |
| White-Yellow | |
| MFCD00043141 | |
| n-succinimidyl 3-maleimidopropionate, 2,5-dioxopyrrolidin-1-yl 3-2,5-dioxo-2,5-dihydro-1h-pyrrol-1-yl propanoate, 3-maleimido propionic acid n-hydroxysuccinimide ester, bmps, 3-maleimidopropionic acid n-succinimidyl ester, 3-maleimidopropionic acid nhs, succinimido 3-maleimidopropanoate, 2,5-dioxopyrrolidin-1-yl 3-2,5-dioxopyrrol-1-yl propanoate, 3-maleimidopropionic acid n-hydroxysuccinimide ester | |
| C1CC(=O)N(C1=O)OC(=O)CCN2C(=O)C=CC2=O | |
| 266.209 | |
| 266.21 | |
| Crystalline Powder |
Chemical Identifiers
| 55750-62-4 | |
| 266.209 | |
| JKHVDAUOODACDU-UHFFFAOYSA-N | |
| 4620597 | |
| C1CC(=O)N(C1=O)OC(=O)CCN2C(=O)C=CC2=O |
| C11H10N2O6 | |
| MFCD00043141 | |
| n-succinimidyl 3-maleimidopropionate, 2,5-dioxopyrrolidin-1-yl 3-2,5-dioxo-2,5-dihydro-1h-pyrrol-1-yl propanoate, 3-maleimido propionic acid n-hydroxysuccinimide ester, bmps, 3-maleimidopropionic acid n-succinimidyl ester, 3-maleimidopropionic acid nhs, succinimido 3-maleimidopropanoate, 2,5-dioxopyrrolidin-1-yl 3-2,5-dioxopyrrol-1-yl propanoate, 3-maleimidopropionic acid n-hydroxysuccinimide ester | |
| (2,5-dioxopyrrolidin-1-yl) 3-(2,5-dioxopyrrol-1-yl)propanoate |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator