missing translation for 'onlineSavingsMsg'
Learn More

N-Succinimidyl 3-Maleimidopropionate 98.0+%, TCI America™

Catalog No. S0427100MG
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S0427100MG 100 mg
S04271G 1 g
S04275G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S0427100MG Supplier TCI America Supplier No. S0427100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 55750-62-4
Molecular Formula C11H10N2O6
Molecular Weight (g/mol) 266.209
MDL Number MFCD00043141
InChI Key JKHVDAUOODACDU-UHFFFAOYSA-N
Synonym n-succinimidyl 3-maleimidopropionate, 2,5-dioxopyrrolidin-1-yl 3-2,5-dioxo-2,5-dihydro-1h-pyrrol-1-yl propanoate, 3-maleimido propionic acid n-hydroxysuccinimide ester, bmps, 3-maleimidopropionic acid n-succinimidyl ester, 3-maleimidopropionic acid nhs, succinimido 3-maleimidopropanoate, 2,5-dioxopyrrolidin-1-yl 3-2,5-dioxopyrrol-1-yl propanoate, 3-maleimidopropionic acid n-hydroxysuccinimide ester
PubChem CID 4620597
IUPAC Name (2,5-dioxopyrrolidin-1-yl) 3-(2,5-dioxopyrrol-1-yl)propanoate
SMILES C1CC(=O)N(C1=O)OC(=O)CCN2C(=O)C=CC2=O

Specifications

Melting Point 168°C
Color White-Yellow
Quantity 100 mg
Formula Weight 266.21
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material N-Succinimidyl 3-Maleimidopropionate [Cross-linking Reagent]
TSCA No
Recommended Storage Refrigerator