missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-sec-Butylpropylamine 98.0+%, TCI America™
Supplier: TCI America B107325ML
Specifications
| N-sec-Butylpropylamine | |
| Yellow | |
| C7H17N | |
| 25 mL | |
| QYNZYUUXSVZDJO-UHFFFAOYSA-N | |
| N-propylbutan-2-amine | |
| 559366 | |
| ≥98.0% (GC) |
| 39190-67-5 | |
| 123°C | |
| MFCD00059398 | |
| 2734 | |
| CCCNC(C)CC | |
| 115.22 | |
| 115.22 | |
| Liquid |
Chemical Identifiers
| 39190-67-5 | |
| 115.22 | |
| QYNZYUUXSVZDJO-UHFFFAOYSA-N | |
| N-propylbutan-2-amine |
| C7H17N | |
| MFCD00059398 | |
| 559366 | |
| CCCNC(C)CC |
Safety and Handling
TSCA : No