missing translation for 'onlineSavingsMsg'
Learn More
Learn More
n-Propyltrimethoxysilane, 98+%
Description
It finds its application as an important material in sol-gel production, provides a hydrophobic surface treatment for inorganic powders or filter materials and it is one of the catalyst component for polyolefin production with Ziegler-Natta catalyst.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It finds its application as an important material in sol-gel production, provides a hydrophobic surface treatment for inorganic powders or filter materials and it is one of the catalyst component for polyolefin production with Ziegler-Natta catalyst.
Solubility
Reacts with water.
Notes
Moisture sensitive. Store away from oxidizing agents and bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
It finds its application as an important material in sol-gel production, provides a hydrophobic surface treatment for inorganic powders or filter materials and it is one of the catalyst component for polyolefin production with Ziegler-Natta catalyst.
Solubility
Reacts with water.
Notes
Moisture sensitive. Store away from oxidizing agents and bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Specifications
| Boiling Point | 142°C to 143°C |
| Quantity | 500 g |
| UN Number | UN1993 |
| Beilstein | 2070458 |
| Flash Point | 34°C (93°F) |
| Solubility Information | Reacts with water. |
| Formula Weight | 164.28 |
| Percent Purity | ≥98% |
| Refractive Index | 1.39 |
| Sensitivity | Moisture sensitive |
| Show More |
Safety and Handling
| Hazard Category | H226-H315-H319 |
| Hazard Statement | GHS H Statement H226-H315-H319-H335 Flammable liquid and vapour. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P210-P233-P240-P241-P242-P243-P264b-P280-P303+P361+P353-P305+P351+P338-P332+P313-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S. |
| EINECSNumber | 213-926-7 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only