missing translation for 'onlineSavingsMsg'
Learn More

N-Propylethylenediamine, 99%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 314750050

Quantity 5g
Packaging Glass bottle

 View more versions of this product

Catalog No. AC314750050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

111-39-7
102.18
CFNHVUGPXZUTRR-UHFFFAOYSA-N
66073
CCCNCCN
C5H14N2
MFCD00008172
n-propylethylenediamine, n-n-propyl ethylenediamine, 1,2-ethanediamine, n-propyl, ethylenediamine, n-propyl, 2-aminoethyl propyl amine, 1,2-ethanediamine, n1-propyl, 2-n-propylamino ethylamine, 2-n-propylaminoethylamine, propylethylenediamine, n-n-propylethylenediamine
N'-propylethane-1,2-diamine

Specifications

N-Propylethylenediamine
111-39-7
100.0
147°C to 150°C
Authentic
Glass bottle
1.4394 to 1.4414
5g
0.82
CFNHVUGPXZUTRR-UHFFFAOYSA-N
N'-propylethane-1,2-diamine
66073
99%
99%
98.5
0.8200g/mL
14°C
99%
C5H14N2
CH3CH2CH2NHCH2CH2NH2
MFCD00008172
n-propylethylenediamine, n-n-propyl ethylenediamine, 1,2-ethanediamine, n-propyl, ethylenediamine, n-propyl, 2-aminoethyl propyl amine, 1,2-ethanediamine, n1-propyl, 2-n-propylamino ethylamine, 2-n-propylaminoethylamine, propylethylenediamine, n-n-propylethylenediamine
CCCNCCN
102.18
102.18
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 203-864-9

SDS
Documents

Documents

Promotions

Promotions