Learn More
n-Propyl propionate, 99%
CAS: 106-36-5 | C6H12O2 | 116.16 g/mol
Supplier: Thermo Scientific Chemicals B20680AK
Description
It has application in automotive refinish, OEM coatings, appliance coatings, printing inks and as polymerization solvent. Its good activity yields efficient resin viscosity reduction that enables the development of lower VOC coatings. Its high electrical resistivity is beneficial in optimizing transfer efficiency in coatings applied via electrostatic spray equipment.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt has application in automotive refinish, OEM coatings, appliance coatings, printing inks and as polymerization solvent. Its good activity yields efficient resin viscosity reduction that enables the development of lower VOC coatings. Its high electrical resistivity is beneficial in optimizing transfer efficiency in coatings applied via electrostatic spray equipment.
Solubility
Soluble in water. (5g/L).
Notes
Store at room temperature. Incompatible with strong oxidizing agents.
Specifications
| n-Propyl propionate | |
| -76°C | |
| 122°C to 124°C | |
| C6H12O2 | |
| CH3CH2CO2CH2CH2CH3 | |
| 250 mL | |
| 14,7867 | |
| Soluble in water. (5g/L). | |
| CCCOC(=O)CC | |
| 116.16 | |
| 116.16 |
| 106-36-5 | |
| 0.881 | |
| 19°C (67°F) | |
| 1.393 | |
| MFCD00009373 | |
| UN3272 | |
| propyl propionate, n-propyl propionate, propanoic acid, propyl ester, n-propyl propanoate, propionic acid, propyl ester, n-propyl n-propionate, propylester kyseliny propionove, unii-g09trv00gk, propyl propionate natural, propionic acid n-propyl ester | |
| MCSINKKTEDDPNK-UHFFFAOYSA-N | |
| propyl propanoate | |
| 7803 | |
| 99% |
Chemical Identifiers
| 106-36-5 | |
| 116.16 | |
| MCSINKKTEDDPNK-UHFFFAOYSA-N | |
| 7803 | |
| CCCOC(=O)CC |
| C6H12O2 | |
| MFCD00009373 | |
| propyl propionate, n-propyl propionate, propanoic acid, propyl ester, n-propyl propanoate, propionic acid, propyl ester, n-propyl n-propionate, propylester kyseliny propionove, unii-g09trv00gk, propyl propionate natural, propionic acid n-propyl ester | |
| propyl propanoate |
Safety and Handling
GHS H Statement
H226-H332
Flammable liquid and vapour.
Harmful if inhaled.
P210-P233-P240-P241-P242-P243-P261-P271-P280-P303+P361+P353-P304+P340-P312-P370+P378q-P501c
H226-H332
DOTInformation : Hazard Class: 3; Packaging Group: III
EINECSNumber : 203-389-7
RTECSNumber : UF7100000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only