missing translation for 'onlineSavingsMsg'
Learn More

n-Propyl bromoacetate, 97%

Catalog No. p-7045776
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1195106 5 g
AAL1195114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1195106 Supplier Thermo Scientific Chemicals Supplier No. L1195106

CAS: 35223-80-4 | C5H9BrO2 | 181.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 35223-80-4
Molecular Formula C5H9BrO2
Molecular Weight (g/mol) 181.03
MDL Number MFCD00075247
InChI Key ISYUCUGTDNJIHV-UHFFFAOYSA-N
Synonym propyl bromoacetate, n-propyl bromoacetate, acetic acid, bromo-, propyl ester, acmc-1ajeq, bromoacetic acid propyl ester, bromo-acetic acid propyl ester, acetic acid, 2-bromo-,propyl ester
PubChem CID 141981
IUPAC Name propyl 2-bromoacetate
SMILES CCCOC(=O)CBr

Specifications

Density 1.399
Boiling Point 175°C to 176°C
Flash Point 82°C (179°F)
Refractive Index 1.451
Quantity 5 g
UN Number UN2922
Beilstein 1745564
Formula Weight 181.04
Percent Purity 97%
Chemical Name or Material n-Propyl bromoacetate
Hazard Category H227-H302+H312+H332-H314-H335
Hazard Statement GHS H Statement
H331-H314-H318-H227-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P210-P235-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only