missing translation for 'onlineSavingsMsg'
Learn More

n-Propyl Alcohol, Reagent grade, 98.0% min., MilliporeSigma™

Catalog No. MPX18153
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MPX18153 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MPX18153 Supplier Technidata Supplier No. PX18153

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 71-23-8
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.10
MDL Number MFCD00002941
InChI Key BDERNNFJNOPAEC-UHFFFAOYSA-N
Synonym 1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcohol
PubChem CID 1031
ChEBI CHEBI:28831
IUPAC Name propan-1-ol
SMILES CCCO

Specifications

Quantity 4 L
Physical Form Liquid
Assay Percent Range 98.0%, min.
Linear Formula CH3CH2CH2OH
Formula Weight 6.1
Grade Reagent
Chemical Name or Material n-Propyl Alcohol
Recommended Storage Room Temperature