missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Propyl Alcohol, Purified (n-Propanol; 1-Propanol), Reagents
Specifications
Specifications
| Melting Point | -127°C |
| Color | Colorless |
| Quantity | 20 L |
| Packaging | Steel Pail |
| Physical Form | Liquid |
| Specific Gravity | 0.8016 |
| Boiling Point | 97°C |
| Grade | Pure |
| Chemical Name or Material | N-Propyl Alcohol, Purified (n-Propanol, 1-Propanol) |