missing translation for 'onlineSavingsMsg'
Learn More

N-Propyl Alcohol, Purified (n-Propanol; 1-Propanol), Reagents

Catalog No. 01337518
Change view
Click to view available options
Quantity:
500 mL
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337518 4 L
01337517 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01337518 Supplier Labchem Inc Supplier No. C21212004C
Only null left

N-Propyl Alcohol, Purified (n-Propanol; 1-Propanol), Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 71-23-8
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.10
InChI Key BDERNNFJNOPAEC-UHFFFAOYSA-N
Synonym n-Propanol, 1-Propyl Alcohol, n-Propyl Alcohol, Propan-1-ol
IUPAC Name propan-1-ol
SMILES CCCO

Specifications

Melting Point -127°C
Color Colorless
Quantity 4 L
Packaging Glass Bottle
Physical Form Liquid
Specific Gravity 0.8016
Boiling Point 97°C
Grade Pure
Chemical Name or Material N-Propyl Alcohol, Purified (n-Propanol, 1-Propanol)