missing translation for 'onlineSavingsMsg'
Learn More

n-Propyl Alcohol, ACS Reagent, for organic synthesis, prep-LC, and general laboratory use, >99.5%, Honeywell

Catalog No. p-7112051
Change view
Click to view available options
Quantity:
4 x 4 L
Packaging:
Glass Bottle
Catalog No. Quantity Packaging
6000108 4 x 4 L Glass Bottle
1 options

Catalog No. 6000108

Supplier: Honeywell Inc LP3224

Only null left

Chemical Identifiers

CAS 71-23-8
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.10
MDL Number MFCD00002941
InChI Key BDERNNFJNOPAEC-UHFFFAOYSA-N
Synonym 1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcohol
PubChem CID 1031
ChEBI CHEBI:28831
IUPAC Name propan-1-ol
SMILES CCCO

Specifications

CAS Max % 100.0000%
Density 0.8053 g/cm3
Boiling Point 97.2°C
Quantity 4 x 4 L
Flash Point 23°C
Packaging Glass Bottle
UN Number UN1274
Beilstein 1098242
Water 0.2% max.
Solubility Information Miscible in Water at 20°C
Formula Weight 60.09
Percent Purity >99.5%
Grade ACS Reagent
Assay by GC Analysis
Chemical Name or Material n-Propyl Alcohol, ACS Reagent, for organic synthesis, prep-LC, and general laboratory use, >99.5%
Show More Show Less
ShelfLife 547 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: III; Proper Shipping Name: n-Propanol