missing translation for 'onlineSavingsMsg'
Learn More

N-Propyl Alcohol, ACS Reagent (n-Propanol; 1-Propanol), Reagents

Catalog No. 01338128
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 L
20 L
4 L
500 mL
Packaging:
Glass Bottle
Poly Bottle
Steel Pail
Catalog No. Quantity Packaging
01338128 20 L Steel Pail
01338129 1 L Glass Bottle
01338127 4 L Poly Bottle
01338126 4 L Glass Bottle
01338125 500 mL Glass Bottle
5 options

Catalog No. 01338128

Supplier: Labchem Inc C112120020E6

Only null left

N-Propyl Alcohol, ACS Reagent (n-Propanol; 1-Propanol), ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 71-23-8
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.10
InChI Key BDERNNFJNOPAEC-UHFFFAOYSA-N
Synonym n-Propanol, 1-Propyl Alcohol, n-Propyl Alcohol, Propan-1-ol
IUPAC Name propan-1-ol
SMILES CCCO

Specifications

Melting Point -127°C
Color Colorless
Boiling Point 97°C
Quantity 20 L
Packaging Steel Pail
Physical Form Liquid
Specific Gravity 0.8016
Grade ACS
Chemical Name or Material N-Propyl Alcohol, ACS Reagent (n-Propanol, 1-Propanol)