missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Propyl Alcohol, ACS Reagent (n-Propanol; 1-Propanol), Reagents
N-Propyl Alcohol, ACS Reagent (n-Propanol; 1-Propanol), ACS, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc C11212004A
Specifications
| N-Propyl Alcohol, ACS Reagent (n-Propanol, 1-Propanol) | |
| 71-23-8 | |
| 97°C | |
| Poly Bottle | |
| 0.8016 | |
| n-Propanol, 1-Propyl Alcohol, n-Propyl Alcohol, Propan-1-ol | |
| CCCO | |
| 60.10 |
| -127°C | |
| Colorless | |
| 4 L | |
| Liquid | |
| C3H8O | |
| BDERNNFJNOPAEC-UHFFFAOYSA-N | |
| propan-1-ol | |
| ACS |
Chemical Identifiers
| 71-23-8 | |
| 60.10 | |
| n-Propanol, 1-Propyl Alcohol, n-Propyl Alcohol, Propan-1-ol | |
| CCCO |
| C3H8O | |
| BDERNNFJNOPAEC-UHFFFAOYSA-N | |
| propan-1-ol |