missing translation for 'onlineSavingsMsg'
Learn More

n-Propyl Alcohol, ACS, 99.5%, Spectrum™ Chemical

Catalog No. 18612046
Change view
Click to view available options
Quantity:
500 mL
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
Drum
Poly Bottle
Steel Can
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18612046 4 L Poly Bottle
18612049 500 mL Amber Glass Bottle
18612045 4 L Amber Glass Bottle
18612047 20 L Steel Can
18612048 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18612046 Supplier Spectrum Chemical Mfg Cor Supplier No. P14304LTPL
Get Up to $500 in Chemicals with Purchase
View all promotions

Chemical Identifiers

CAS 71-23-8
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.10
InChI Key BDERNNFJNOPAEC-UHFFFAOYSA-N
IUPAC Name propan-1-ol
SMILES CCCO

Specifications

CAS Max % 1
Packaging Poly Bottle
Quantity 4 L
Residue after Evaporation 1e-05
Percent Purity 0.995
Grade ACS
Titratable Acid 0.0004 meq/g

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.