missing translation for 'onlineSavingsMsg'
Learn More

n-Propyl acrylate, 95%, stab. with 0.1% 4-methoxyphenol

Catalog No. p-7045183
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0971409 10 g
AAL0971418 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0971409 Supplier Thermo Scientific Chemicals Supplier No. L0971409
Only null left

CAS: 925-60-0 | C6H10O2 | 114.14 g/mol

n-propyl acrylate is used in coatings, textile and leather finishes, paper coatings, for adhesives, for acrylic fibers. It is used as a monomer for emulsion polymers and solution polymers in latex paints.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
n-propyl acrylate is used in coatings, textile and leather finishes, paper coatings, for adhesives, for acrylic fibers. It is used as a monomer for emulsion polymers and solution polymers in latex paints.

Solubility
Immiscible with water.

Notes
Incompatible with oxidizing agents.

Chemical Identifiers

CAS 925-60-0
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.14
MDL Number MFCD00048693
InChI Key PNXMTCDJUBJHQJ-UHFFFAOYSA-N
Synonym n-propyl acrylate, propyl acrylate, 2-propenoic acid, propyl ester, propyl 2-propenoate, acrylic acid, propyl ester, propylacrylate, ccris 8584, 1-propyl acrylate, acrylic acid propyl, acrylic acid propyl ester
PubChem CID 13550
IUPAC Name propyl prop-2-enoate
SMILES CCCOC(=O)C=C

Specifications

Density 0.92
Boiling Point 43°C to 44°C (40 mmHg)
Flash Point 24°C (75°F)
Refractive Index 1.4123
Quantity 10 g
UN Number UN3272
Beilstein 1744954
Solubility Information Immiscible with water.
Formula Weight 114.15
Percent Purity 95%
Chemical Name or Material n-Propyl acrylate, stabilized with 0.1% 4-methoxyphenol
Show More Show Less
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 213-120-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only