missing translation for 'onlineSavingsMsg'
Learn More

N-(Propargyloxy)phthalimide 98.0+%, TCI America™

Catalog No. P23421G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P23421G 1 g
P23425G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P23421G Supplier TCI America Supplier No. P23421G

Chemical Identifiers

CAS 4616-63-1
Molecular Formula C11H7NO3
Molecular Weight (g/mol) 201.181
MDL Number MFCD00005890
InChI Key HBGZBVPXPDNXOV-UHFFFAOYSA-N
Synonym n-propargyloxy phthalimide, n-prop-2-ynyloxy phthalimide, 2-prop-2-yn-1-yloxy isoindoline-1,3-dione, n-propargyloxyphthalimide, 2-prop-2-yn-1-yloxy isoindole-1,3-dione, 2-prop-2-ynyloxybenzo c azoline-1,3-dione, acmc-20aka8, n-2-propynoxyphthalimide, n-2-propynyloxy phthalimide, phthalimide, n-2-propynyloxy
PubChem CID 78357
IUPAC Name 2-prop-2-ynoxyisoindole-1,3-dione
SMILES C#CCON1C(=O)C2=CC=CC=C2C1=O

Specifications

Melting Point 140°C
Color White-Yellow
Quantity 1 g
Formula Weight 201.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N-(Propargyloxy)phthalimide