missing translation for 'onlineSavingsMsg'
Learn More

N-(Propargyloxy)phthalimide, 98%, Thermo Scientific™

$176.52 - $332.26

Chemical Identifiers

CAS 4616-63-1
Molecular Formula C11H7NO3
Molecular Weight (g/mol) 201.18
MDL Number MFCD00005890
InChI Key HBGZBVPXPDNXOV-UHFFFAOYSA-N
Synonym n-propargyloxy phthalimide, n-prop-2-ynyloxy phthalimide, 2-prop-2-yn-1-yloxy isoindoline-1,3-dione, n-propargyloxyphthalimide, 2-prop-2-yn-1-yloxy isoindole-1,3-dione, 2-prop-2-ynyloxybenzo c azoline-1,3-dione, acmc-20aka8, n-2-propynoxyphthalimide, n-2-propynyloxy phthalimide, phthalimide, n-2-propynyloxy
PubChem CID 78357
IUPAC Name 2-prop-2-ynoxyisoindole-1,3-dione
SMILES C#CCON1C(=O)C2=CC=CC=C2C1=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC131490100
SDS View Documents
Thermo Scientific Chemicals
131490100
10 g Glass bottle N/A
Only null left
 
AC131490250
SDS View Documents
Thermo Scientific Chemicals
131490250
25 g Glass bottle N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

4616-63-1
201.18
HBGZBVPXPDNXOV-UHFFFAOYSA-N
78357
C#CCON1C(=O)C2=CC=CC=C2C1=O
C11H7NO3
MFCD00005890
n-propargyloxy phthalimide, n-prop-2-ynyloxy phthalimide, 2-prop-2-yn-1-yloxy isoindoline-1,3-dione, n-propargyloxyphthalimide, 2-prop-2-yn-1-yloxy isoindole-1,3-dione, 2-prop-2-ynyloxybenzo c azoline-1,3-dione, acmc-20aka8, n-2-propynoxyphthalimide, n-2-propynyloxy phthalimide, phthalimide, n-2-propynyloxy
2-prop-2-ynoxyisoindole-1,3-dione

Specifications

4616-63-1
100.0
Beige to Brown
98%
C11H7NO3
10 g
HBGZBVPXPDNXOV-UHFFFAOYSA-N
2-prop-2-ynoxyisoindole-1,3-dione
78357
98%
N-(Propargyloxy)phthalimide, 98%
97.5
149°C to 152°C
Authentic
Glass bottle
MFCD00005890
n-propargyloxy phthalimide, n-prop-2-ynyloxy phthalimide, 2-prop-2-yn-1-yloxy isoindoline-1,3-dione, n-propargyloxyphthalimide, 2-prop-2-yn-1-yloxy isoindole-1,3-dione, 2-prop-2-ynyloxybenzo c azoline-1,3-dione, acmc-20aka8, n-2-propynoxyphthalimide, n-2-propynyloxy phthalimide, phthalimide, n-2-propynyloxy
C#CCON1C(=O)C2=CC=CC=C2C1=O
201.18
201.18
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 225-024-0

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only