missing translation for 'onlineSavingsMsg'
Learn More

N-Piperidineethanol, 99%

Catalog No. p-185375
Change view
Click to view available options
Quantity:
5 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC152110050 5 g Glass bottle
AC152111000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC152110050 Supplier Thermo Scientific Chemicals Supplier No. 152110050

CAS: 3040-44-6 | C7H15NO | 129.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3040-44-6
Molecular Formula C7H15NO
Molecular Weight (g/mol) 129.2
MDL Number MFCD00006512
InChI Key KZTWONRVIPPDKH-UHFFFAOYSA-N
Synonym 2-piperidinoethanol, 1-piperidineethanol, 2-piperidin-1-yl ethanol, 1-2-hydroxyethyl piperidine, n-2-hydroxyethyl piperidine, n-piperidineethanol, 2-1-piperidinyl ethanol, n-hydroxyethyl piperidine, beta-piperidylethanol, beta-piperidinoethanol
PubChem CID 18232
ChEBI CHEBI:61238
IUPAC Name 2-piperidin-1-ylethanol
SMILES C1CCN(CC1)CCO

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 16.7°C
Color Brown
Density 0.9730g/mL
Boiling Point 198°C to 203°C
Flash Point 77°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4794 to 1.4814
Quantity 5 g
Beilstein 20, 25
Specific Gravity 0.973
Solubility Information Solubility in water: miscible
Viscosity 20.9 mPa.s (20°C)
Formula Weight 129.2
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material N-Piperidineethanol, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF SWALLOWED
EINECSNumber 221-244-6
RTECSNumber TM8052000
TSCA TSCA

RUO – Research Use Only