missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Phthaloylglycine 98.0+%, TCI America™
$31.68 - $335.52
Chemical Identifiers
| CAS | 4702-13-0 |
|---|---|
| Molecular Formula | C10H7NO4 |
| Molecular Weight (g/mol) | 205.169 |
| MDL Number | MFCD00005900 |
| InChI Key | WQINSVOOIJDOLJ-UHFFFAOYSA-N |
| Synonym | n-phthaloylglycine, phthalimidoacetic acid, n-phthalylglycine, phthaloylglycine, n-carboxymethyl phthalimide, n,n-phthaloylglycine, n-phthalylglycin, 1,3-dioxo-2-isoindolineacetic acid, 1,3-dioxo-1,3-dihydro-2h-isoindol-2-yl acetic acid, 2-1,3-dioxoisoindolin-2-yl acetic acid |
| PubChem CID | 20825 |
| IUPAC Name | 2-(1,3-dioxoisoindol-2-yl)acetic acid |
| SMILES | C1=CC=C2C(=C1)C(=O)N(C2=O)CC(=O)O |
Chemical Identifiers
| 4702-13-0 | |
| 205.169 | |
| WQINSVOOIJDOLJ-UHFFFAOYSA-N | |
| 20825 | |
| C1=CC=C2C(=C1)C(=O)N(C2=O)CC(=O)O |
| C10H7NO4 | |
| MFCD00005900 | |
| n-phthaloylglycine, phthalimidoacetic acid, n-phthalylglycine, phthaloylglycine, n-carboxymethyl phthalimide, n,n-phthaloylglycine, n-phthalylglycin, 1,3-dioxo-2-isoindolineacetic acid, 1,3-dioxo-1,3-dihydro-2h-isoindol-2-yl acetic acid, 2-1,3-dioxoisoindolin-2-yl acetic acid | |
| 2-(1,3-dioxoisoindol-2-yl)acetic acid |
Specifications
| 4702-13-0 | |
| White | |
| MFCD00005900 | |
| n-phthaloylglycine, phthalimidoacetic acid, n-phthalylglycine, phthaloylglycine, n-carboxymethyl phthalimide, n,n-phthaloylglycine, n-phthalylglycin, 1,3-dioxo-2-isoindolineacetic acid, 1,3-dioxo-1,3-dihydro-2h-isoindol-2-yl acetic acid, 2-1,3-dioxoisoindolin-2-yl acetic acid | |
| C1=CC=C2C(=C1)C(=O)N(C2=O)CC(=O)O | |
| 205.169 | |
| 205.17 | |
| Crystalline Powder |
| 196°C | |
| C10H7NO4 | |
| 25 g | |
| WQINSVOOIJDOLJ-UHFFFAOYSA-N | |
| 2-(1,3-dioxoisoindol-2-yl)acetic acid | |
| 20825 | |
| ≥98.0% (HPLC,T) | |
| N-Phthaloylglycine |
Safety and Handling
RTECSNumber : NR3439000
TSCA : No