missing translation for 'onlineSavingsMsg'
Learn More

N-Phthaloylglycine 98.0+%, TCI America™

Catalog No. P096325G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P096325G 25 g
P0963500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P096325G Supplier TCI America Supplier No. P096325G
Only null left

Chemical Identifiers

CAS 4702-13-0
Molecular Formula C10H7NO4
Molecular Weight (g/mol) 205.169
MDL Number MFCD00005900
InChI Key WQINSVOOIJDOLJ-UHFFFAOYSA-N
Synonym n-phthaloylglycine, phthalimidoacetic acid, n-phthalylglycine, phthaloylglycine, n-carboxymethyl phthalimide, n,n-phthaloylglycine, n-phthalylglycin, 1,3-dioxo-2-isoindolineacetic acid, 1,3-dioxo-1,3-dihydro-2h-isoindol-2-yl acetic acid, 2-1,3-dioxoisoindolin-2-yl acetic acid
PubChem CID 20825
IUPAC Name 2-(1,3-dioxoisoindol-2-yl)acetic acid
SMILES C1=CC=C2C(=C1)C(=O)N(C2=O)CC(=O)O

Specifications

Melting Point 196°C
Color White
Quantity 25 g
Formula Weight 205.17
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-Phthaloylglycine
RTECSNumber NR3439000
TSCA No