missing translation for 'onlineSavingsMsg'
Learn More

N-Phthaloylglycine, 98+%

Catalog No. AAA1592018
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1592018 50 g
AAA1592030 250 g
AAA159200B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1592018 Supplier Thermo Scientific Chemicals Supplier No. A1592018
Only null left

CAS: 4702-13-0 | C10H7NO4 | 205.169 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4702-13-0
Molecular Formula C10H7NO4
Molecular Weight (g/mol) 205.169
MDL Number MFCD00005900
InChI Key WQINSVOOIJDOLJ-UHFFFAOYSA-N
Synonym n-phthaloylglycine, phthalimidoacetic acid, n-phthalylglycine, phthaloylglycine, n-carboxymethyl phthalimide, n,n-phthaloylglycine, n-phthalylglycin, 1,3-dioxo-2-isoindolineacetic acid, 1,3-dioxo-1,3-dihydro-2h-isoindol-2-yl acetic acid, 2-1,3-dioxoisoindolin-2-yl acetic acid
PubChem CID 20825
IUPAC Name 2-(1,3-dioxoisoindol-2-yl)acetic acid
SMILES C1=CC=C2C(=C1)C(=O)N(C2=O)CC(=O)O

Specifications

Melting Point 194°C to 198°C
Beilstein 184174
Quantity 50 g
Formula Weight 205.17
Percent Purity ≥98%
Chemical Name or Material N-Phthaloylglycine
Hazard Category H302-H315-H319-H335-H500
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 225-177-3
RTECSNumber NR3439000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only