missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Phthaloyl-L-glutamic Anhydride 98.0+%, TCI America™
$93.21 - $275.87
Chemical Identifiers
| CAS | 25830-77-7 |
|---|---|
| Molecular Formula | C13H9NO5 |
| Molecular Weight (g/mol) | 259.217 |
| MDL Number | MFCD00067324 |
| InChI Key | ICDLEMPZXFCQEB-VIFPVBQESA-N |
| PubChem CID | 11807393 |
| IUPAC Name | 2-[(3S)-2,6-dioxooxan-3-yl]isoindole-1,3-dione |
| SMILES | C1CC(=O)OC(=O)C1N2C(=O)C3=CC=CC=C3C2=O |
Chemical Identifiers
| 25830-77-7 | |
| 259.217 | |
| ICDLEMPZXFCQEB-VIFPVBQESA-N | |
| 2-[(3S)-2,6-dioxooxan-3-yl]isoindole-1,3-dione |
| C13H9NO5 | |
| MFCD00067324 | |
| 11807393 | |
| C1CC(=O)OC(=O)C1N2C(=O)C3=CC=CC=C3C2=O |
Specifications
| 25830-77-7 | |
| White | |
| MFCD00067324 | |
| ICDLEMPZXFCQEB-VIFPVBQESA-N | |
| 2-[(3S)-2,6-dioxooxan-3-yl]isoindole-1,3-dione | |
| 11807393 | |
| ≥98.0% (T) | |
| N-Phthaloyl-L-glutamic Anhydride |
| 205°C | |
| C13H9NO5 | |
| 1 g | |
| C1CC(=O)OC(=O)C1N2C(=O)C3=CC=CC=C3C2=O | |
| 259.217 | |
| 259.22 | |
| Crystalline Powder |
Safety and Handling
TSCA : No