missing translation for 'onlineSavingsMsg'
Learn More

N-Phenylmaleimide 98.0+%, TCI America™

Catalog No. p-7134835
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P090025G 25 g
P0900100G 100 g
P0900500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P090025G Supplier TCI America Supplier No. P090025G
Only null left

Chemical Identifiers

CAS 941-69-5
Molecular Formula C10H7NO2
Molecular Weight (g/mol) 173.171
MDL Number MFCD00005502
InChI Key HIDBROSJWZYGSZ-UHFFFAOYSA-N
Synonym n-phenylmaleimide, maleanil, maleinanil, maleimidobenzene, 1h-pyrrole-2,5-dione, 1-phenyl, 1-phenyl-1h-pyrrole-2,5-dione, maleimide, n-phenyl, 1-phenyl-2,5-dihydro-1h-pyrrole-2,5-dione, unii-9u9kt462vw, n-fenylimid kyseliny maleinove
PubChem CID 13662
IUPAC Name 1-phenylpyrrole-2,5-dione
SMILES C1=CC=C(C=C1)N2C(=O)C=CC2=O

Specifications

Melting Point 90°C
Color Yellow
Boiling Point 181°C
Quantity 25 g
UN Number 2811
Formula Weight 173.17
Percent Purity ≥98.0% (GC)
Physical Form Flakes
Chemical Name or Material N-Phenylmaleimide
EINECSNumber (5)-5727
RTECSNumber ON5950000
TSCA Yes