missing translation for 'onlineSavingsMsg'
Learn More

N-Phenylmaleimide, 98+%

Catalog No. p-7022034
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1461614 25 g
AAA1461622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1461614 Supplier Thermo Scientific Chemicals Supplier No. A1461614

CAS: 941-69-5 | C10H7NO2 | 173.171 g/mol

N-Phenylmaleimide is used as a dipolarophile in 1,3-cycloaddition reaction with a nitrone, useful for the preparation of crystalline adducts of dienes. It is used as a plastic modifier.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Phenylmaleimide is used as a dipolarophile in 1,3-cycloaddition reaction with a nitrone, useful for the preparation of crystalline adducts of dienes. It is used as a plastic modifier.

Solubility
Soluble in water (slightly ), methanol, ethanol, and benzene.

Notes
Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.

Chemical Identifiers

CAS 941-69-5
Molecular Formula C10H7NO2
Molecular Weight (g/mol) 173.171
MDL Number MFCD00005502
InChI Key HIDBROSJWZYGSZ-UHFFFAOYSA-N
Synonym n-phenylmaleimide, maleanil, maleinanil, maleimidobenzene, 1h-pyrrole-2,5-dione, 1-phenyl, 1-phenyl-1h-pyrrole-2,5-dione, maleimide, n-phenyl, 1-phenyl-2,5-dihydro-1h-pyrrole-2,5-dione, unii-9u9kt462vw, n-fenylimid kyseliny maleinove
PubChem CID 13662
IUPAC Name 1-phenylpyrrole-2,5-dione
SMILES C1=CC=C(C=C1)N2C(=O)C=CC2=O

Specifications

Melting Point 87°C to 91°C
Boiling Point 162°C to 163°C (12 mmHg)
Quantity 25 g
UN Number UN2811
Beilstein 125098
Merck Index 14,7299
Solubility Information Soluble in water (slightly ),methanol,ethanol,and benzene.
Formula Weight 173.17
Percent Purity ≥98%
Chemical Name or Material N-Phenylmaleimide
Hazard Category H301-H315-H319
Hazard Statement GHS H Statement
H301-H319
Toxic if swallowed.
Causes serious eye irritation.
Precautionary Statement P264b-P270-P280-P301+P310-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 213-382-0
RTECSNumber ON5950000
TSCA Yes

RUO – Research Use Only