missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Phenylglycine Ethyl Ester 98.0+%, TCI America™
Supplier: TCI America P178125G
Specifications
| N-Phenylglycine Ethyl Ester | |
| 56°C | |
| 140°C | |
| MFCD00038315 | |
| n-phenylglycine ethyl ester, ethyl phenylglycinate, ethyl 2-phenylamino acetate, ethyl n-phenylglycinate, glycine, n-phenyl-, ethyl ester, unii-7xl9t8bk0u, ethyl anilinoacetate, n-phenylglycineethylester, n-phenylglycine ethyl, 7xl9t8bk0u | |
| CCOC(=O)CNC1=CC=CC=C1 | |
| 179.219 | |
| 179.22 | |
| Crystalline Powder |
| 2216-92-4 | |
| White-Yellow | |
| C10H13NO2 | |
| 25 g | |
| MLSGRWDEDYJNER-UHFFFAOYSA-N | |
| ethyl 2-anilinoacetate | |
| 75190 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 2216-92-4 | |
| 179.219 | |
| MLSGRWDEDYJNER-UHFFFAOYSA-N | |
| 75190 | |
| CCOC(=O)CNC1=CC=CC=C1 |
| C10H13NO2 | |
| MFCD00038315 | |
| n-phenylglycine ethyl ester, ethyl phenylglycinate, ethyl 2-phenylamino acetate, ethyl n-phenylglycinate, glycine, n-phenyl-, ethyl ester, unii-7xl9t8bk0u, ethyl anilinoacetate, n-phenylglycineethylester, n-phenylglycine ethyl, 7xl9t8bk0u | |
| ethyl 2-anilinoacetate |
Safety and Handling
TSCA : Yes