missing translation for 'onlineSavingsMsg'
Learn More

N-Phenylglycine Ethyl Ester 98.0+%, TCI America™

Catalog No. P17815G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P17815G 5 g
P178125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P17815G Supplier TCI America Supplier No. P17815G

Chemical Identifiers

CAS 2216-92-4
Molecular Formula C10H13NO2
Molecular Weight (g/mol) 179.219
MDL Number MFCD00038315
InChI Key MLSGRWDEDYJNER-UHFFFAOYSA-N
Synonym n-phenylglycine ethyl ester, ethyl phenylglycinate, ethyl 2-phenylamino acetate, ethyl n-phenylglycinate, glycine, n-phenyl-, ethyl ester, unii-7xl9t8bk0u, ethyl anilinoacetate, n-phenylglycineethylester, n-phenylglycine ethyl, 7xl9t8bk0u
PubChem CID 75190
IUPAC Name ethyl 2-anilinoacetate
SMILES CCOC(=O)CNC1=CC=CC=C1

Specifications

Melting Point 56°C
Color White-Yellow
Boiling Point 140°C
Quantity 5 g
Formula Weight 179.22
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N-Phenylglycine Ethyl Ester