missing translation for 'onlineSavingsMsg'
Learn More

N-Phenylglycine ethyl ester, 99%

Catalog No. AAB2228209
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2228209 10 g
AAB2228218 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2228209 Supplier Thermo Scientific Chemicals Supplier No. B2228209

CAS: 2216-92-4 | C10H13NO2 | 179.219 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2216-92-4
Molecular Formula C10H13NO2
Molecular Weight (g/mol) 179.219
MDL Number MFCD00038315
InChI Key MLSGRWDEDYJNER-UHFFFAOYSA-N
Synonym n-phenylglycine ethyl ester, ethyl phenylglycinate, ethyl 2-phenylamino acetate, ethyl n-phenylglycinate, glycine, n-phenyl-, ethyl ester, unii-7xl9t8bk0u, ethyl anilinoacetate, n-phenylglycineethylester, n-phenylglycine ethyl, 7xl9t8bk0u
PubChem CID 75190
IUPAC Name ethyl 2-anilinoacetate
SMILES CCOC(=O)CNC1=CC=CC=C1

Specifications

Melting Point 55°C to 58°C
Merck Index 14,7292
Quantity 10 g
Formula Weight 179.22
Percent Purity 99%
Chemical Name or Material N-Phenylglycine ethyl ester
EINECSNumber 218-702-2
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only