missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N-Phenylglycine, 97%

Catalog No. AAA1418218
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1418218 50 g
AAA1418209 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1418218 Supplier Thermo Scientific Chemicals Supplier No. A1418218
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 103-01-5 | C8H8NNaO2 | 173.15 g/mol

N-Phenylglycine coupled with Glycidyl methacrylate to create a surface active comonomer that promotes adhesive bonding of materials to hard tooth tissues.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Phenylglycine coupled with Glycidyl methacrylate to create a surface active comonomer that promotes adhesive bonding of materials to hard tooth tissues.

Solubility
Soluble in water.

Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.

Chemical Identifiers

CAS 103-01-5
Molecular Formula C8H8NNaO2
Molecular Weight (g/mol) 173.15
MDL Number MFCD00014009
InChI Key YEMGQZDWLLBIEY-UHFFFAOYSA-M
Synonym n-phenylglycine, anilinoacetic acid, 2-phenylamino acetic acid, glycine, n-phenyl, glycine, phenyl, n-phenylglycin, n-phenylaminoacetic acid, phenylamino acetic acid, h-phenylgly-oh, n-phenyl glycine
PubChem CID 66025
ChEBI CHEBI:55477
IUPAC Name 2-anilinoacetic acid
SMILES [Na+].[O-]C(=O)CNC1=CC=CC=C1

Specifications

Melting Point 117°C to 119°C
Beilstein 509838
Merck Index 14,7292
Solubility Information Soluble in water.
Quantity 50 g
Formula Weight 151.17
Percent Purity 97%
Chemical Name or Material N-Phenylglycine
EINECSNumber 203-070-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only