missing translation for 'onlineSavingsMsg'
Learn More

N-Phenylglycine 97.0+%, TCI America™

Catalog No. P018025G
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P018025G 25 g
P0180100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P018025G Supplier TCI America Supplier No. P018025G

Chemical Identifiers

CAS 103-01-5
Molecular Formula C8H8NNaO2
Molecular Weight (g/mol) 173.15
MDL Number MFCD00014009
InChI Key YEMGQZDWLLBIEY-UHFFFAOYSA-M
Synonym n-phenylglycine, anilinoacetic acid, 2-phenylamino acetic acid, glycine, n-phenyl, glycine, phenyl, n-phenylglycin, n-phenylaminoacetic acid, phenylamino acetic acid, h-phenylgly-oh, n-phenyl glycine
PubChem CID 66025
ChEBI CHEBI:55477
IUPAC Name sodium 2-(phenylamino)acetate
SMILES [Na+].[O-]C(=O)CNC1=CC=CC=C1

Specifications

Melting Point 125°C
Color White-Yellow
Quantity 25 g
Formula Weight 151.17
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Phenylglycine
EINECSNumber (3)-1441
TSCA Yes