missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ N-Phenylglycine, 93%
Chemical Identifiers
| CAS | 103-01-5 |
|---|---|
| Molecular Formula | C8H8NNaO2 |
| Molecular Weight (g/mol) | 173.15 |
| MDL Number | MFCD00014009 |
| InChI Key | YEMGQZDWLLBIEY-UHFFFAOYSA-M |
| Synonym | n-phenylglycine, anilinoacetic acid, 2-phenylamino acetic acid, glycine, n-phenyl, glycine, phenyl, n-phenylglycin, n-phenylaminoacetic acid, phenylamino acetic acid, h-phenylgly-oh, n-phenyl glycine |
| PubChem CID | 66025 |
| ChEBI | CHEBI:55477 |
| IUPAC Name | 2-anilinoacetic acid |
| SMILES | [Na+].[O-]C(=O)CNC1=CC=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC276080250
|
Thermo Scientific™
276080250 |
25g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC276081000
|
Thermo Scientific™
276081000 |
100g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 103-01-5 | |
| 173.15 | |
| YEMGQZDWLLBIEY-UHFFFAOYSA-M | |
| 66025 | |
| 2-anilinoacetic acid |
| C8H8NNaO2 | |
| MFCD00014009 | |
| n-phenylglycine, anilinoacetic acid, 2-phenylamino acetic acid, glycine, n-phenyl, glycine, phenyl, n-phenylglycin, n-phenylaminoacetic acid, phenylamino acetic acid, h-phenylgly-oh, n-phenyl glycine | |
| CHEBI:55477 | |
| [Na+].[O-]C(=O)CNC1=CC=CC=C1 |
Specifications
| 103-01-5 | |
| 100.0 | |
| 91% min. (HPLC) | |
| C8H8NNaO2 | |
| MFCD00014009 | |
| 12, 468 | |
| n-phenylglycine, anilinoacetic acid, 2-phenylamino acetic acid, glycine, n-phenyl, glycine, phenyl, n-phenylglycin, n-phenylaminoacetic acid, phenylamino acetic acid, h-phenylgly-oh, n-phenyl glycine | |
| YEMGQZDWLLBIEY-UHFFFAOYSA-M | |
| 2-anilinoacetic acid | |
| 66025 | |
| 151.17 | |
| N-Phenylglycine, 93% |
| 91.0 | |
| Authentic | |
| Glass bottle | |
| C6H5NHCH2CO2H | |
| 25g | |
| 15, 7403 | |
| Solubility in water: moderately soluble. Other solubilities: sparingly soluble in ether | |
| [Na+].[O-]C(=O)CNC1=CC=CC=C1 | |
| 173.15 | |
| CHEBI:55477 | |
| 93% |
Safety and Handling
EINECSNumber : 203-070-2
TSCA : TSCA
RUO â Research Use Only