missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N-Phenylglycine, 93%

Catalog No. AC276081000
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC276081000 100 g Glass bottle
AC276080250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC276081000 Supplier Thermo Scientific Chemicals Supplier No. 276081000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 103-01-5 | C8H8NNaO2 | 173.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 103-01-5
Molecular Formula C8H8NNaO2
Molecular Weight (g/mol) 173.15
MDL Number MFCD00014009
InChI Key YEMGQZDWLLBIEY-UHFFFAOYSA-M
Synonym n-phenylglycine, anilinoacetic acid, 2-phenylamino acetic acid, glycine, n-phenyl, glycine, phenyl, n-phenylglycin, n-phenylaminoacetic acid, phenylamino acetic acid, h-phenylgly-oh, n-phenyl glycine
PubChem CID 66025
ChEBI CHEBI:55477
IUPAC Name 2-anilinoacetic acid
SMILES [Na+].[O-]C(=O)CNC1=CC=CC=C1

Specifications

CAS Min % 91.0
CAS Max % 100.0
Melting Point 121.0°C to 127.0°C
Color Brown-Yellow to Ochre
Infrared Spectrum Authentic
Assay Percent Range 91% min. (HPLC)
Packaging Glass bottle
Linear Formula C6H5NHCH2CO2H
Quantity 100 g
Beilstein 12, 468
Merck Index 15, 7403
Solubility Information Solubility in water: moderately soluble. Other solubilities: sparingly soluble in ether
Formula Weight 151.17
Percent Purity 93%
Physical Form Fine Powder
Chemical Name or Material N-Phenylglycine, 93%
Show More Show Less
EINECSNumber 203-070-2
TSCA TSCA

RUO – Research Use Only