missing translation for 'onlineSavingsMsg'
Learn More

N-Phenylethylenediamine 98.0+%, TCI America™

Catalog No. P110110G
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P110110G 10 g
P110125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P110110G Supplier TCI America Supplier No. P110110G

Chemical Identifiers

CAS 1664-40-0
Molecular Formula C8H12N2
Molecular Weight (g/mol) 136.198
MDL Number MFCD00008162
InChI Key OCIDXARMXNJACB-UHFFFAOYSA-N
Synonym n-phenylethylenediamine, n1-phenylethane-1,2-diamine, n-2-aminoethyl aniline, 1,2-ethanediamine, n-phenyl, ethylenediamine, n-phenyl, n-phenyl-1,2-ethanediamine, benzenamine, n-2-aminoethyl, n-phenylethane-1,2-diamine, 1,2-ethanediamine, n1-phenyl, 2-aminoethyl phenylamine
PubChem CID 74270
IUPAC Name N'-phenylethane-1,2-diamine
SMILES C1=CC=C(C=C1)NCCN

Specifications

Color Yellow
Boiling Point 263°C
Quantity 10 g
Formula Weight 136.20
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material N-Phenylethylenediamine