missing translation for 'onlineSavingsMsg'
Learn More

N-Phenylethylenediamine, 99%

Catalog No. p-3758670
Change view
Click to view available options
Quantity:
10 mL
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC130610100 10 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC130610100 Supplier Thermo Scientific Chemicals Supplier No. 130610100

CAS: 1664-40-0 | C8H12N2 | 136.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1664-40-0
Molecular Formula C8H12N2
Molecular Weight (g/mol) 136.2
MDL Number MFCD00008162
InChI Key OCIDXARMXNJACB-UHFFFAOYSA-N
Synonym n-phenylethylenediamine, n1-phenylethane-1,2-diamine, n-2-aminoethyl aniline, 1,2-ethanediamine, n-phenyl, ethylenediamine, n-phenyl, n-phenyl-1,2-ethanediamine, benzenamine, n-2-aminoethyl, n-phenylethane-1,2-diamine, 1,2-ethanediamine, n1-phenyl, 2-aminoethyl phenylamine
PubChem CID 74270
IUPAC Name N'-phenylethane-1,2-diamine
SMILES C1=CC=C(C=C1)NCCN

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Yellow
Density 1.0400g/mL
Boiling Point 262.0°C to 264.0°C
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Refractive Index 1.5855 to 1.5875
Linear Formula C6H5NHCH2CH2NH2
Quantity 10 mL
Beilstein 12, 543
Specific Gravity 1.04
Formula Weight 136.2
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material N-Phenylethylenediamine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 216-773-4

RUO – Research Use Only