missing translation for 'onlineSavingsMsg'
Learn More

N-Phenylbenzylamine 98.0+%, TCI America™

Catalog No. p-7134457
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P015925G 25 g
P0159100G 100 g
P0159500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P015925G Supplier TCI America Supplier No. P015925G
Only null left

Chemical Identifiers

CAS 103-32-2
Molecular Formula C13H13N
Molecular Weight (g/mol) 183.254
MDL Number MFCD00003018
InChI Key GTWJETSWSUWSEJ-UHFFFAOYSA-N
Synonym n-phenylbenzylamine, benzylaniline, benzenemethanamine, n-phenyl, phenylbenzylamine, benzylphenylamine, n-monobenzylaniline, aniline, n-benzyl, benzylamine, n-phenyl, benzenamine, n-phenylmethyl, n-benzyl aniline
PubChem CID 66028
IUPAC Name N-benzylaniline
SMILES C1=CC=C(C=C1)CNC2=CC=CC=C2

Specifications

Melting Point 37°C
Color White-Yellow
Boiling Point 307°C
Quantity 25 g
Formula Weight 183.25
Percent Purity ≥98.0% (GC)
Physical Form Crystals
Chemical Name or Material N-Phenylbenzylamine
EINECSNumber (3)-0374
TSCA Yes