missing translation for 'onlineSavingsMsg'
Learn More

N-Phenylbenzylamine, 99%

CAS: 103-32-2 | C13H13N | 183.25 g/mol

$335.70 - $335.70

Chemical Identifiers

CAS 103-32-2
Molecular Formula C13H13N
Molecular Weight (g/mol) 183.25
MDL Number MFCD00003018
InChI Key GTWJETSWSUWSEJ-UHFFFAOYSA-N
Synonym n-phenylbenzylamine, benzylaniline, benzenemethanamine, n-phenyl, phenylbenzylamine, benzylphenylamine, n-monobenzylaniline, aniline, n-benzyl, benzylamine, n-phenyl, benzenamine, n-phenylmethyl, n-benzyl aniline
PubChem CID 66028
IUPAC Name N-benzylaniline
SMILES C1=CC=C(C=C1)CNC2=CC=CC=C2
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC417391000
SDS View Documents
Thermo Scientific Chemicals
417391000
100 g Glass bottle
Each for $335.70
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

103-32-2
183.25
GTWJETSWSUWSEJ-UHFFFAOYSA-N
66028
C1=CC=C(C=C1)CNC2=CC=CC=C2
C13H13N
MFCD00003018
n-phenylbenzylamine, benzylaniline, benzenemethanamine, n-phenyl, phenylbenzylamine, benzylphenylamine, n-monobenzylaniline, aniline, n-benzyl, benzylamine, n-phenyl, benzenamine, n-phenylmethyl, n-benzyl aniline
N-benzylaniline

Specifications

103-32-2
100.0
Beige-Yellow to Colorless
306°C to 307°C
Authentic
Glass bottle
C6H5CH2NHC6H5
100 g
1.06
n-phenylbenzylamine, benzylaniline, benzenemethanamine, n-phenyl, phenylbenzylamine, benzylphenylamine, n-monobenzylaniline, aniline, n-benzyl, benzylamine, n-phenyl, benzenamine, n-phenylmethyl, n-benzyl aniline
GTWJETSWSUWSEJ-UHFFFAOYSA-N
N-benzylaniline
66028
99%
N-Phenylbenzylamine
98.5
34°C to 38°C
1.0600g/mL
152°C
98.5% min. (GC)
C13H13N
MFCD00003018
12, 1023
15, 1129
Solubility in water: insoluble in water. Other solubilities: very soluble in ethanol,ether,soluble in chloroform
C1=CC=C(C=C1)CNC2=CC=CC=C2
183.25
183.25
Crystalline Powder or Low Melting Mass
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 203-100-4

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only