missing translation for 'onlineSavingsMsg'
Learn More

N-Phenylbenzylamine, 99%

Catalog No. AC417390050
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC417390050 5 g Glass bottle
AC417390250 25 g Glass bottle
AC417391000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC417390050 Supplier Thermo Scientific Chemicals Supplier No. 417390050
Only null left

CAS: 103-32-2 | C13H13N | 183.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 103-32-2
Molecular Formula C13H13N
Molecular Weight (g/mol) 183.25
MDL Number MFCD00003018
InChI Key GTWJETSWSUWSEJ-UHFFFAOYSA-N
Synonym n-phenylbenzylamine, benzylaniline, benzenemethanamine, n-phenyl, phenylbenzylamine, benzylphenylamine, n-monobenzylaniline, aniline, n-benzyl, benzylamine, n-phenyl, benzenamine, n-phenylmethyl, n-benzyl aniline
PubChem CID 66028
IUPAC Name N-benzylaniline
SMILES C1=CC=C(C=C1)CNC2=CC=CC=C2

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 34°C to 38°C
Color Beige-Yellow to Colorless
Density 1.0600g/mL
Boiling Point 306°C to 307°C
Flash Point 152°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H5CH2NHC6H5
Quantity 5 g
Beilstein 12, 1023
Specific Gravity 1.06
Merck Index 15, 1129
Solubility Information Solubility in water: insoluble in water. Other solubilities: very soluble in ethanol,ether,soluble in chloroform
Formula Weight 183.25
Percent Purity 99%
Physical Form Crystalline Powder or Low Melting Mass
Chemical Name or Material N-Phenylbenzylamine
Show More Show Less
EINECSNumber 203-100-4

RUO – Research Use Only