missing translation for 'onlineSavingsMsg'
Learn More

N-Phenylbenzamidine, 97%

Catalog No. AAA1321914
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1321914 25 g
AAA1321906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1321914 Supplier Thermo Scientific Chemicals Supplier No. A1321914
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1527-91-9 | C13H12N2 | 196.253 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1527-91-9
Molecular Formula C13H12N2
Molecular Weight (g/mol) 196.253
MDL Number MFCD00019730
InChI Key MPYOKHFSBKUKPQ-UHFFFAOYSA-N
Synonym n-phenylbenzamidine, n-phenylbenzenecarboximidamide, n'-phenylbenzamidine, n1-phenylbenzene-1-carboximidamide, n2-phenylbenzamidine, n-phenylbenzimidamide, acmc-20ak1f, maybridge1_004766, iminophenylmethyl phenylamine, n-phenyl-benzenecarboximidamide
PubChem CID 200127
IUPAC Name N'-phenylbenzenecarboximidamide
SMILES C1=CC=C(C=C1)C(=NC2=CC=CC=C2)N

Specifications

Melting Point 117°C to 120°C
Quantity 25 g
Beilstein 608471
Sensitivity Hygroscopic
Formula Weight 196.25
Percent Purity 97%
Chemical Name or Material N-Phenylbenzamidine
Hazard Category H302+H312+H332-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only