missing translation for 'onlineSavingsMsg'
Learn More

N-Phenylaza-15-crown 5-Ether 96.0+%, TCI America™

Catalog No. P11435G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P11435G 5 g
P11431G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P11435G Supplier TCI America Supplier No. P11435G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 66750-10-5 | Formula: C16H25NO4 | Weight: 295.379 g/mol

Chemical Identifiers

CAS 66750-10-5
Molecular Formula C16H25NO4
Molecular Weight (g/mol) 295.379
MDL Number MFCD00040513
InChI Key SGDQOAKAHLFKBV-UHFFFAOYSA-N
Synonym n-phenylaza-15-crown 5-ether, n-phenylaza-15-crown-5, 1,4,7,10-tetraoxa-13-azacyclopentadecane, 13-phenyl, sgdqoakahlfkbv-uhfffaoysa, n-phenyl-13-aza-1,4,7,10-tetraoxacyclopentadecane, n-phenyl-4,7,10,13-tetraoxa-1-aza-cyclopentadecane, 13-phenyl-1,4,7,10-tetraoxa-13-aza-cyclopentadecane
PubChem CID 626432
IUPAC Name 13-phenyl-1,4,7,10-tetraoxa-13-azacyclopentadecane
SMILES C1COCCOCCOCCOCCN1C2=CC=CC=C2

Specifications

Melting Point 46°C
Color Red-White
Quantity 5 g
Formula Weight 295.38
Percent Purity ≥96.0% (GC,T)
Grade Not Available
Physical Form Crystalline Powder
Chemical Name or Material N-Phenylaza-15-crown 5-Ether