missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Phenylaza-15-crown 5-Ether 96.0+%, TCI America™
Specifications
Specifications
| Melting Point | 46°C |
| Color | Red-White |
| Quantity | 1 g |
| Formula Weight | 295.38 |
| Percent Purity | ≥96.0% (GC,T) |
| Grade | Not Available |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | N-Phenylaza-15-crown 5-Ether |
Safety and Handling
| TSCA | No |