missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Phenyl-p-phenylenediamine, 98%
Description
p-Anilinoaniline is an antioxidant that has shown to cause a reduction of NOx emissions from soybean biodiesel powered DI diesel engines but has increased CO and HC emissions. It is also used to produce dyes, pharmaceuticals, and photographic chemicals and frmerly used as a hair dye.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
p-Anilinoaniline is an antioxidant that has shown to cause a reduction of NOx emissions from soybean biodiesel powered DI diesel engines but has increased CO and HC emissions. It is also used to produce dyes, pharmaceuticals, and photographic chemicals and frmerly used as a hair dye.
Solubility
Soluble in water at 20°C 0.6g/L. Soluble in (10 mg/mL) ethanol.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
p-Anilinoaniline is an antioxidant that has shown to cause a reduction of NOx emissions from soybean biodiesel powered DI diesel engines but has increased CO and HC emissions. It is also used to produce dyes, pharmaceuticals, and photographic chemicals and frmerly used as a hair dye.
Solubility
Soluble in water at 20°C 0.6g/L. Soluble in (10 mg/mL) ethanol.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Specifications
| Melting Point | 72°C to 76°C |
| Density | 1.09 |
| Boiling Point | 354°C |
| Flash Point | 193°C (379°F) |
| Quantity | 1000 g |
| UN Number | UN3077 |
| Beilstein | 908935 |
| Solubility Information | Soluble in water at 20°C 0.6g/L. Soluble in (10mg/mL) ethanol. |
| Formula Weight | 184.24 |
| Percent Purity | 98% |
| Show More |
Safety and Handling
| Hazard Category | H302+H332-H315-H317-H319-H335 |
| Hazard Statement | GHS H Statement H301-H315-H319-H317-H335 Toxic if swallowed. Causes skin irritation. Causes serious eye irritation. May cause an allergic skin reaction. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P333+P313-P362-P501c |
| DOTInformation | Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S. |
| EINECSNumber | 202-951-9 |
| RTECSNumber | ST3150000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only