missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Phenyl-p-phenylenediamine, 98%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| CAS Min % | 97.5 |
| CAS Max % | 100.0 |
| Melting Point | 69.0°C to 75.0°C |
| Boiling Point | 354.0°C |
| Flash Point | >250°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 97.5% min. (GC) |
| Packaging | Glass bottle |
| Linear Formula | C6H5NHC6H4NH2 |
| Quantity | 100 g |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement May cause respiratory irritation. Causes skin irritation. Causes serious eye irritation. Harmful if inhaled. May cause an allergic skin reaction. Harmful if swallowed. Very toxic to aquatic life |
| Precautionary Statement | GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. IF INHALED: If breathing is difficult,remove to fresh air and keep at rest in a position comfortable for breathing. If experiencing respiratory symptoms: Call a POISON CENT |
| EINECSNumber | 202-951-9 |
| RTECSNumber | ST3150000 |
| TSCA | TSCA |
RUO – Research Use Only