missing translation for 'onlineSavingsMsg'
Learn More

N-Phenyl-2-naphthylamine 97.0+%, TCI America™

Catalog No. p-7134481
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P019825G 25 g
P0198500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P019825G Supplier TCI America Supplier No. P019825G
Only null left

Chemical Identifiers

CAS 135-88-6
Molecular Formula C16H13N
Molecular Weight (g/mol) 219.29
MDL Number MFCD00004052
InChI Key KEQFTVQCIQJIQW-UHFFFAOYSA-N
Synonym n-phenyl-2-naphthylamine, n-2-naphthyl aniline, neozone, aceto pbn, stabilizator ar, agerite powder, anilinonaphthalene, antioxidant 116, pbna, 2-naphthylphenylamine
PubChem CID 8679
ChEBI CHEBI:34877
IUPAC Name N-phenylnaphthalen-2-amine
SMILES N(C1=CC=CC=C1)C1=CC=C2C=CC=CC2=C1

Specifications

Melting Point 108°C
Boiling Point 237°C
Quantity 25 g
UN Number 3077
Formula Weight 219.29
Percent Purity ≥97.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material N-Phenyl-2-naphthylamine
EINECSNumber (4)-0330
RTECSNumber QM4550000
TSCA Yes