missing translation for 'onlineSavingsMsg'
Learn More

N-Phenyl-2-naphthylamine, 97%

Catalog No. AC439910250
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC439910250 25 g Glass bottle
AC439911000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC439910250 Supplier Thermo Scientific Chemicals Supplier No. 439910250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 135-88-6 | C16H13N | 219.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 135-88-6
Molecular Formula C16H13N
Molecular Weight (g/mol) 219.29
MDL Number MFCD00004052
InChI Key KEQFTVQCIQJIQW-UHFFFAOYSA-N
Synonym n-phenyl-2-naphthylamine, n-2-naphthyl aniline, neozone, aceto pbn, stabilizator ar, agerite powder, anilinonaphthalene, antioxidant 116, pbna, 2-naphthylphenylamine
PubChem CID 8679
ChEBI CHEBI:34877
IUPAC Name N-phenylnaphthalen-2-amine
SMILES N(C1=CC=CC=C1)C1=CC=C2C=CC=CC2=C1

Specifications

Melting Point 105°C to 108°C
Boiling Point 395°C
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 25 g
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol,ether,benzene,acetone,acetic acid
Formula Weight 219.28
Percent Purity 97%
Chemical Name or Material N-Phenyl-2-naphthylamine
Hazard Category GHS Signal Word: Warning
EINECSNumber 205-223-9

RUO – Research Use Only