missing translation for 'onlineSavingsMsg'
Learn More

N-Phenyl-1-naphthylamine 98.0+%, TCI America™

Catalog No. P019725G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P019725G 25 g
P0197500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P019725G Supplier TCI America Supplier No. P019725G

Chemical Identifiers

CAS 90-30-2
Molecular Formula C16H13N
Molecular Weight (g/mol) 219.287
MDL Number MFCD00003878
InChI Key XQVWYOYUZDUNRW-UHFFFAOYSA-N
Synonym n-phenyl-1-naphthylamine, 1-anilinonaphthalene, pana, antioxidant pan, n-1-naphthyl aniline, neozone a, 1-naphthalenamine, n-phenyl, phenylnaphthylamine, nonox a, aceto pan
PubChem CID 7013
ChEBI CHEBI:34876
IUPAC Name N-phenylnaphthalen-1-amine
SMILES C1=CC=C(C=C1)NC2=CC=CC3=CC=CC=C32

Specifications

Melting Point 60°C
Boiling Point 200°C
Quantity 25 g
UN Number 3077
Formula Weight 219.29
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N-Phenyl-1-naphthylamine
EINECSNumber (4)-0329
RTECSNumber QM4500000
TSCA Yes