missing translation for 'onlineSavingsMsg'
Learn More

n-Pentylbenzene, 96%

Catalog No. AAA1573406
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1573406 5 g
AAA1573414 25 g
AAA1573422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1573406 Supplier Thermo Scientific Chemicals Supplier No. A1573406
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 538-68-1 | C11H16 | 148.249 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 538-68-1
Molecular Formula C11H16
Molecular Weight (g/mol) 148.249
MDL Number MFCD00009502
InChI Key PWATWSYOIIXYMA-UHFFFAOYSA-N
Synonym 1-phenylpentane, n-pentylbenzene, amylbenzene, n-amylbenzene, benzene, pentyl, 1-pentylbenzene, 1-phenyl-n-pentane, pentane, 1-phenyl, pentyl-benzene, pentane, phenyl
PubChem CID 10864
IUPAC Name pentylbenzene
SMILES CCCCCC1=CC=CC=C1

Specifications

Melting Point -75°C
Density 0.863
Boiling Point 204°C to 205°C
Flash Point 65°C (149°F)
Refractive Index 1.488
Quantity 5 g
UN Number UN3082
Beilstein 1904359
Merck Index 14,600
Formula Weight 148.25
Percent Purity 96%
Chemical Name or Material n-Pentylbenzene
Show More Show Less
Hazard Category H227
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
EINECSNumber 208-701-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only