missing translation for 'onlineSavingsMsg'
Learn More

n-Pentane, Reagents

Catalog No. p-200021035
Change view
Click to view available options
Quantity:
500 mL
4 L
20 L
200 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337149 200 L
01337148 20 L
01337147 4 L
01337146 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 01337149 Supplier Labchem Inc Supplier No. C2265600200L

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

n-Pentane, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 109-66-0
Molecular Formula C5H12
Molecular Weight (g/mol) 72.15
InChI Key OFBQJSOFQDEBGM-UHFFFAOYSA-N
Synonym n-Pentane, normal Pentane
IUPAC Name pentane
SMILES CCCCC

Specifications

Melting Point -130°C
Color Colorless
Boiling Point 36°C
Quantity 200 L
Packaging Steel Drum
Physical Form Liquid
Specific Gravity 0.6264
Grade Reagent
Chemical Name or Material n-Pentane