missing translation for 'onlineSavingsMsg'
Learn More
Learn More
n-Pentane, Reagents
Specifications
Specifications
| Melting Point | -130°C |
| Color | Colorless |
| Quantity | 20 L |
| Packaging | Steel Pail |
| Physical Form | Liquid |
| Specific Gravity | 0.6264 |
| Boiling Point | 36°C |
| Grade | Reagent |
| Chemical Name or Material | n-Pentane |